missing translation for 'onlineSavingsMsg'
Learn More

Intelsius™ PathoPouch™ 95kPa with Absorbent

The PathoPouch™ 95kPa with Absorbent is a flexible, leak-proof secondary pouch designed to withstand 95 kPa of pressure in compliance with Category A and B regulations. Suitable for the transportation of a variety of sample containers.

Brand:  Intelsius™ BP001

Encompass_Preferred

 View more versions of this product

Product Code. 17404602

  • POA / Pack of 50

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Suitable for the transportation of a variety of samples including:
  • Diagnostic Samples
  • Category A and B Infectious Substances
  • Blood Vials
  • Swabs
Specifications

Specifications

6 Bays
A4
Leak-proof Sample Pouch
308 x 230 mm (12.1 x 9.1 in.)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Intelsius™ PathoPouch™ 95kPa with Absorbent

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.