missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Phosphate Buffered Saline Tablets (PBS)
Phosphate Buffered Saline Tablets (PBS) are a balanced salt mixture for easy PBS buffer preparation for a variety of applications, including reagent preparation, diluting cells for flow cytometry, and cell culture reagent.
Brand: MP Biomedicals™ 092810305
193.80 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
This item is not returnable.
View return policy
Description
- No need for weighing and mixing chemicals
- Simply dissolve PBS tablets in water
- One tablet dissolved in 100mL water yields buffer containing 0.01M phosphate, 0.0027M KCl, and 0.137M NaCl, pH 7.4 at 25°C
- Research use only

Specifications
No Calcium, No Magnesium | |
100 Tablets | |
MFCD00011360 | |
[Cl-].[K+] | |
74.55 |
Phosphate Buffered Saline (PBS) | |
ClK | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium chloride | |
Tablet |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Phosphate Buffered Saline Tablets (PBS) > Phosphate Buffered Saline Tablets (PBS), 100 Tablets
Spot an opportunity for improvement?Share a Content Correction