Learn More
Perfluoro-1-butanesulfonyl fluoride, 92%, Thermo Scientific Chemicals
29.15 € - 233.16 €
Chemical Identifiers
CAS | 375-72-4 |
---|---|
Molecular Formula | C4F10O2S |
Molecular Weight (g/mol) | 302.09 |
MDL Number | MFCD00007422 |
InChI Key | LUYQYZLEHLTPBH-UHFFFAOYSA-N |
Synonym | nonafluorobutanesulfonyl fluoride, perfluorobutanesulfonyl fluoride, perfluoro-1-butanesulfonyl fluoride, perfluorobutanesulfonylfluoride, perfluorobutylsulfonyl fluoride, nonafluorobutanesulphonyl fluoride, perfluoro n-butylsulfonyl fluoride, nonafluorobutane-1-sulfonyl fluoride, nonafluoro-1-butanesulfonyl fluoride, 1-butanesulfonyl fluoride, 1,1,2,2,3,3,4,4,4-nonafluoro |
PubChem CID | 67814 |
IUPAC Name | 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonyl fluoride |
SMILES | C(C(C(F)(F)S(=O)(=O)F)(F)F)(C(F)(F)F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10668444
|
Thermo Scientific Acros
312340050 |
5 g | Glass bottle |
29.15 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10349410
|
Thermo Scientific Acros
312340250 |
25 g | Glass bottle |
82.81 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10540224
|
Thermo Scientific Acros
312341000 |
100 g | Glass bottle |
233.16 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
375-72-4 | |
302.09 | |
LUYQYZLEHLTPBH-UHFFFAOYSA-N | |
67814 | |
C(C(C(F)(F)S(=O)(=O)F)(F)F)(C(F)(F)F)(F)F |
C4F10O2S | |
MFCD00007422 | |
nonafluorobutanesulfonyl fluoride, perfluorobutanesulfonyl fluoride, perfluoro-1-butanesulfonyl fluoride, perfluorobutanesulfonylfluoride, perfluorobutylsulfonyl fluoride, nonafluorobutanesulphonyl fluoride, perfluoro n-butylsulfonyl fluoride, nonafluorobutane-1-sulfonyl fluoride, nonafluoro-1-butanesulfonyl fluoride, 1-butanesulfonyl fluoride, 1,1,2,2,3,3,4,4,4-nonafluoro | |
1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonyl fluoride |
Specifications
375-72-4 | |
1.6800g/mL | |
Authentic | |
C4F10O2S | |
1.3 | |
1.68 | |
LUYQYZLEHLTPBH-UHFFFAOYSA-N | |
1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonyl fluoride | |
67814 | |
92% | |
Perfluoro-1-butanesulfonyl fluoride, 92% |
Colorless to Yellow | |
64°C | |
90% min. (GC) | |
CF3(CF2)3SO2F | |
MFCD00007422 | |
nonafluorobutanesulfonyl fluoride, perfluorobutanesulfonyl fluoride, perfluoro-1-butanesulfonyl fluoride, perfluorobutanesulfonylfluoride, perfluorobutylsulfonyl fluoride, nonafluorobutanesulphonyl fluoride, perfluoro n-butylsulfonyl fluoride, nonafluorobutane-1-sulfonyl fluoride, nonafluoro-1-butanesulfonyl fluoride, 1-butanesulfonyl fluoride, 1,1,2,2,3,3,4,4,4-nonafluoro | |
C(C(C(F)(F)S(=O)(=O)F)(F)F)(C(F)(F)F)(F)F | |
302.09 | |
302.09 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-792-6
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.