missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ pH Indicator Paper Strips

Dip the pH test paper into the solution to be tested and compare the colour with the colour chart provided. Fisherbrand™ pH Indicator Paper Strips provide quick and easy pH determination.

Brand:  Fisherbrand™ PH010S

 View more versions of this product

Product Code. 15575669

This item is not returnable. View return policy

Description

Description

  • Includes color chart label for easy reference and pH level determination
  • 200 strips
Specifications

Specifications

0 to 11
Color indicator chart
pH Test Strip
Strip
200/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ pH Indicator Paper Strips > pH Range: 0 to 11

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.