Learn More
Macherey-Nagel™ pH-Fix Test Strips in PT Tubes
Provides quick and easy pH value testing at the point of interest. Macherey Nagel™ pH-Fix Test Strips in PT Tubes don't need any calibration, set-up or maintenance.
Brand:  Macherey-Nagel™ 92111
Product Code. 10025504
  • 16.64 € / Pack of 100
Edge
Explore more special offers
Description
Description
Provides quick and easy pH value testing at the point of interest. Macherey Nagel™ pH-Fix Test Strips in PT Tubes don't need any calibration, set-up or maintenance.
  • Long plastic handle protects against toxic, dangerous or aggressive substances
  • Features up to 4 different indicator pads that cover the whole pH range
  • No bleeding due to color bounded indicator dyes
  • Tube features simple one-hand operation, prevents against jamming, and is virtually unbreakable
Specifications
Specifications
0 to 14
100 Pack
pH Test Strip
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ pH-Fix Test Strips in PT Tubes > pH: 0 to 14