Learn More
Fisherbrand™ Phadebas™ Amylase Test Tablets (50)
Determine Α-amylase activitiy for applications in fields including the food industry, biotech, and detergents. Fisherbrand™ Phadebas™ Amylase Test Tablets come with instructions and a batch standard curve.
Brand:  Fisherbrand™ 1301
Product Code. 12865983
  • 306.00 € / 50 tablets
Edge
Explore more special offers
Description
Description
  • For a wide variety of applications in fields including the food industry, biotech, and detergents.
  • This test has been on the market for over 35 years and is well documented in the scientific literature.
  • Includes instructions for use in English, French, Spanish, and German.
  • Includes a batch standard curve for quantitative assessment.
Specifications
Specifications
Determining Α-amylase activity
50 Tablets
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Phadebas™ Amylase Test Tablets (50) > 50 Tablets