missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phalloidin Labeling Probes

Achieve precise and reliable F-actin staining with fluorescent and biotinylated phalloidins. Phalloidin conjugates are widely used in imaging applications to selectively label F-actin in a variety of sample types including fixed and permeabilized cells, tissue sections, and cell-free experiments.

Brand:  Invitrogen™ F432

597.36 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.



 View more versions of this product

Product Code. 10615963

  • 896.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Green
496/516 nm
Fluorescence Microscope, Flow Cytometer, Confocal Microscope, Compatible with FITC/GFP filter set
Room Temperature
Phalloidin
Store in freezer -5°C to -30°C and protect from light.
FITC (Fluorescein)
300 Units
Classic Dyes
Actin, Cytoskeleton
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3011741 Certificate TypeCertificate of Analysis Date07/10/2024 Catalog NumberF432
Lot Number2663857 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog NumberF432
Lot Number2611662 Certificate TypeCertificate of Analysis Date13/06/2023 Catalog NumberF432
Lot Number2515909 Certificate TypeCertificate of Analysis Date22/09/2022 Catalog NumberF432
Lot Number2406491 Certificate TypeCertificate of Analysis Date27/09/2021 Catalog NumberF432
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Phalloidin Labeling Probes > Color: Green, Excitation Wavelength Range: 496/516 nm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.