missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein solution, Indicator, Reag. Ph. Eur., 1% in ethanol, Honeywell Fluka™

Indicator, Reag. Ph. Eur., 1% in ethanol

55.20 € - 2445.97 €

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
PubChem CID 4764
ChEBI CHEBI:34914
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15698370
View Documents
Honeywell-Fluka
34607-30ML
30 mL
55.20 €
30mL
Please to purchase this item. Need a web account? Register with us today!
15614310
View Documents
Honeywell-Fluka
34607-500ML
500 mL
59.48 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15604310
View Documents
Honeywell-Fluka
34607-1L
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15634310
View Documents
Honeywell-Fluka
34607-6X1L
6 x 1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15624310
View Documents
Honeywell-Fluka
34607-50L
50 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

77-09-8
318.33
KJFMBFZCATUALV-UHFFFAOYSA-N
CHEBI:34914
C20H14O4
MFCD00005913
4764
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

77-09-8
MFCD00005913
284423
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
318.33
CHEBI:34914
14°C (57.2°F)
C20H14O4
UN1170
KJFMBFZCATUALV-UHFFFAOYSA-N
3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
4764
318.32g/mol
Phenolphthalein solution
Videos
Safety and Handling

Safety and Handling

P201-P210-P280-P308 + P313-P337 + P313-P403 + P235

Product Identifier
  • Phenolphthalein solution

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : phenolphthalein, ethanol; ethyl alcohol

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.