Learn More
Phenyl vinyl sulfone, 99+%, Thermo Scientific Chemicals
Description
Phenyl vinyl sulfone is used as building blocks in the synthesis of a number of organic substances, participate in polymerization reactions, nucleophilic addition reaction, and in cycloaddition reactions. It is also used in the used in the synthesis of reactive dyes, synthesis of textile fibers, in particular cellulose fibers such as cotton and linen. Other applications in the field of medicine includes as powerful inhibitors of certain enzymatic processes. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phenyl vinyl sulfone is used as building blocks in the synthesis of a number of organic substances, participate in polymerization reactions, nucleophilic addition reaction, and in cycloaddition reactions. It is also used in the used in the synthesis of reactive dyes, synthesis of textile fibers, in particular cellulose fibers such as cotton and linen. Other applications in the field of medicine includes as powerful inhibitors of certain enzymatic processes. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.
Solubility
It is soluble in most common organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Specifications
Specifications
Melting Point | 67°C to 71°C |
Quantity | 1 g |
Beilstein | 1906894 |
Solubility Information | It is soluble in most common organic solvents. |
Formula Weight | 168.22 |
Percent Purity | ≥99% |
Chemical Name or Material | Phenyl vinyl sulfone |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10173749 | Certificate of Analysis | 22/11/2021 | A14794.03,A14794.14,A14794.06 |
10214312 | Certificate of Analysis | 22/11/2021 | A14794.03,A14794.14,A14794.06 |
I8742A | Certificate of Analysis | 22/11/2021 | A14794.03,A14794.14,A14794.06 |
L19S051 | Certificate of Analysis | 22/11/2021 | A14794.03,A14794.14,A14794.06 |
10179701 | Certificate of Analysis | 22/11/2021 | A14794.03,A14794.14,A14794.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.