Learn More
Thermo Scientific™ Phire Plant Direct PCR Master Mix
Thermo Scientific Phire Plant Direct PCR Master Mix is designed to amplify DNA directly from plant samples.
165.00 € - 652.00 €
Specifications
GC-Rich PCR Performance | Medium |
---|---|
PCR Method | Direct PCR |
Polymerase | Phire Hot Start II DNA Polymerase |
Reaction Speed | Fast |
Product Type | Direct PCR Master Mix |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15255156
|
Thermo Scientific™
F160S |
250 Reactions |
165.00 €
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15265156
|
Thermo Scientific™
F160L |
1250 Reactions |
652.00 €
1250 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific Phire Plant Direct PCR Master Mix is designed to amplify DNA directly from plant samples. No DNA purification is required prior to PCR. The kit is based on Phire Hot Start II DNA Polymerase, a specially engineered enzyme with a DNA binding domain. The unique features of this DNA polymerase make it extremely robust and tolerant of many PCR inhibitors present in plant material like complex polysaccharides, polyphenols and others.
Phire Plant Direct PCR Master Mix has been tested with leaves and seeds from a wide variety of plant species as well as fungi, bacteria and yeast. The kit includes a complete set of optimized reagents and detailed protocols making it an ideal choice for amplification of plant DNA.
Highlights
- Direct PCR—sample is added directly to PCR reaction, therefore there is no need for time-consuming and expensive DNA purification steps.
- Dilution & Storage protocol available—allows tens of PCR reactions from one tiny sample and allows re-testing
- Specially engineered Phire Hot Start II DNA polymerase—extremely short PCR protocol times
- Master Mix format with premixed gel loading dye—minimizes possibility of cross-contamination, reduces sample handling and allows direct loading on gel
Applications
- Genotyping
- Transgene detection
- Knockout analysis
- Sequencing
The optimal annealing temperature for Phire DNA Polymerases may differ significantly from that of Taq-based polymerases.
For optimal results start by accurately calculating your Tm with our Tm calculator.
Specifications
Medium | |
Phire Hot Start II DNA Polymerase | |
Direct PCR Master Mix | |
Genotyping, Hot-start PCR | |
Built-In Hot Start | |
SuperMix or Master Mix | |
DNA |
Direct PCR | |
Fast | |
Wet or Dry Ice | |
2X | |
Blunt | |
Plant |
For Research Use Only. Not for use in diagnostic procedures.