missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phire Plant Direct PCR Master Mix

Thermo Scientific Phire Plant Direct PCR Master Mix is designed to amplify DNA directly from plant samples.

Brand:  Thermo Scientific™ F160S

107.25 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15255156

  • 165.00 € / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Phire Plant Direct PCR Master Mix is designed to amplify DNA directly from plant samples. No DNA purification is required prior to PCR. The kit is based on Phire Hot Start II DNA Polymerase, a specially engineered enzyme with a DNA binding domain. The unique features of this DNA polymerase make it extremely robust and tolerant of many PCR inhibitors present in plant material like complex polysaccharides, polyphenols, and others.

Phire Plant Direct PCR Master Mix has been tested with leaves and seeds from a wide variety of plant species as well as fungi, bacteria, and yeast. The kit includes a complete set of optimized reagents and detailed protocols, making it an ideal choice for amplification of plant DNA.

Features

  • Direct PCR allows sample to be added directly to PCR reaction, reducing time-consuming and expensive DNA purification steps
  • Available Dilution & Storage protocol allows several PCR reactions from one sample
  • Specially engineered Phire Hot Start II DNA Polymerase for extremely short PCR protocol times
  • Master mix format with premixed gel loading dye minimizes risk of cross-contamination, reduces sample handling, and allows direct loading on gel

Applications

  • Genotyping
  • Transgene detection
  • Knockout analysis
  • Sequencing

Notes

  • Annealing temperature for Phire DNA Polymerases may differ significantly from that of Taq-based polymerases.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 2X Phire Plant Direct PCR Master Mix, 2 x 1.25 mL
• Dilution Buffer, 1 x 5 mL
• 25 μM Control Primer Mix, 40 μL
• Nuclease-free water, 2 x 1.25 mL
• O'GeneRuler Express DNA Ladder, 50 μg (sufficient for 100 applications)

Store at -20°C.
Direct PCR
Fast
Direct PCR Master Mix
Wet or Dry Ice
2X
250 Reactions
SuperMix or Master Mix
DNA
Medium
Phire Hot Start II DNA Polymerase
Yes
250 reactions
Genotyping, Hot-start PCR
Built-In Hot Start
Blunt
Plant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.