missing translation for 'onlineSavingsMsg'
Learn More
Phloroglucinol Dihydrate, 99%, Thermo Scientific Chemicals Product Code.: 10775431

Phloroglucinol Dihydrate, 99%, Thermo Scientific Chemicals

Product Code. 10775431
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10775431

Brand: Thermo Scientific Acros 131040250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6099-90-7
Molecular Formula C6H6O3·2H2O
Molecular Weight (g/mol) 162.14
MDL Number MFCD00149090
InChI Key MPYXTIHPALVENR-UHFFFAOYSA-N
Synonym phloroglucinol dihydrate, benzene-1,3,5-triol dihydrate, 1,3,5-trihydroxybenzene dihydrate, 1,3,5-benzenetriol, dihydrate, phloroglucinol, dihydrate, sym-trihydroxybenze, unii-0x7b22brg1, ccris 6521, 1,3,5-benzoltrioldihydrat, phloroglucinoldihydrateShow MoreShow Less
PubChem CID 80196
IUPAC Name benzene-1,3,5-triol;dihydrate
SMILES C1=C(C=C(C=C1O)O)O.O.O

Specifications

Melting Point 218.0°C to 221.0°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H3(OH)3·2H2O
Quantity 25 g
Beilstein 06, 1092
Merck Index 15, 7439
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and pyridine
Formula Weight 162.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Phloroglucinol dihydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0435506 Certificate TypeCertificate of Analysis Date15/04/2024 Catalog Number131041000,131040250,131045000
Lot NumberA0425873 Certificate TypeCertificate of Analysis Date25/12/2023 Catalog Number131041000,131040250,131045000
Lot NumberA0455703 Certificate TypeCertificate of Analysis Date28/08/2023 Catalog Number131041000,131040250,131045000
Lot NumberA0455706 Certificate TypeCertificate of Analysis Date28/08/2023 Catalog Number131041000,131040250,131045000
Lot NumberA0455311 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number131041000,131040250,131045000
compliance-icons
Product Identifier
  • Phloroglucinol dihydrate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phloroglucinol Dihydrate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.