Learn More
Phosphoric acid, 85% aq. soln., Thermo Scientific Chemicals
Description
Phosphoric acid is used mainly in fertilizers. It is used as an external standard for phosphorus-31 Nuclear magnetic resonance (NMR); as an electrolyte in phosphoric acid fuel cells and as a catalyst in the hydration of alkenes to produce alcohols, predominantly ethanol. It finds application as a buffer agent in biology and chemistry; as a solution for anodizing and as a cleaner by construction trades to remove mineral deposits, cementitious smears and hard water stains. It is also used as a pH adjuster in cosmetics and skin-care products and a dispersing agent in detergents and leather treatment. It plays an important role in the preparation of albumin derivatives. It is an important starting material used in the preparation of disodium hydrogen phosphate, trisodium phosphate, calcium dihydrogen phosphate and monohydrogen phosphate. It is also used in dentistry and orthodontics as an etching solution as well as a sanitizing agent in dairy, food and brewing industries. It serves as an acidic fruitlike flavoring in food products. It is widely used to remove rust in steel tools or surfaces by converting the rusted ion in to water soluble phosphate compound.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phosphoric acid is used mainly in fertilizers. It is used as an external standard for phosphorus-31 Nuclear magnetic resonance (NMR); as an electrolyte in phosphoric acid fuel cells and as a catalyst in the hydration of alkenes to produce alcohols, predominantly ethanol. It finds application as a buffer agent in biology and chemistry; as a solution for anodizing and as a cleaner by construction trades to remove mineral deposits, cementitious smears and hard water stains. It is also used as a pH adjuster in cosmetics and skin-care products and a dispersing agent in detergents and leather treatment. It plays an important role in the preparation of albumin derivatives. It is an important starting material used in the preparation of disodium hydrogen phosphate, trisodium phosphate, calcium dihydrogen phosphate and monohydrogen phosphate. It is also used in dentistry and orthodontics as an etching solution as well as a sanitizing agent in dairy, food and brewing industries. It serves as an acidic fruitlike flavoring in food products. It is widely used to remove rust in steel tools or surfaces by converting the rusted ion in to water soluble phosphate compound.
Notes
Incompatible with strong bases and powdered metals.

Specifications
Specifications
Melting Point | 19°C to 21°C |
Concentration | 85% aq. soln. |
Density | 1.7 |
Quantity | 500 mL |
Linear Formula | H3PO4 |
UN Number | UN1805 |
Merck Index | 14,7344 |
Solubility Information | Miscible with water,ethanol and mixture of ether: alcohol (3:1). |
Formula Weight | 98 |
Chemical Name or Material | Phosphoric acid |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N14L025 | Certificate of Analysis | 20/02/2025 | A18067.AP,A18067.0F,A18067.0D |
X15K001 | Certificate of Analysis | 25/10/2024 | A18067.AP,A18067.0F,A18067.0D |
Q17K039 | Certificate of Analysis | 23/04/2024 | A18067.AP,A18067.0F,A18067.0D |
N23K019 | Certificate of Analysis | 05/03/2024 | A18067.AP,A18067.0F,A18067.0D |
Y01H075 | Certificate of Analysis | 20/02/2024 | A18067.AP,A18067.0F,A18067.0D |
Safety and Handling

- Phosphoric acid, 85% aqueous solution
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Orthophosphoric acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.