Learn More
Palintest™ Photometer Tablet Reagents
Photometer reagents in convenient tablet form. Palintest™; Photmoeter Tablet Reagents are available for a variety of water testing needs.
Brand: Palintest™ PM188
This item is not returnable.
View return policy
Description
- Convenient tablet form.
- Easy to use.
- Available for a variety of water testing needs.
Specifications
50 | |
0 to 500mg/L | |
Alkalinity Total (Alkaphot) Test Kit |
50/Pk. | |
50 Tests |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Palintest™ Photometer Tablet Reagents > Range: 0 to 500mg/L
Spot an opportunity for improvement?Share a Content Correction