missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion Blood Direct PCR Master Mix

Thermo Scientific Phusion Blood Direct PCR Master Mix is designed for amplification of DNA from whole blood. It eliminates the need for a separate DNA purification step prior to PCR.

119.00 € - 514.00 €

Specifications

GC-Rich PCR Performance High
PCR Method Direct PCR
Polymerase Phusion Blood II DNA Polymerase
Reaction Speed Fast
Includes Label or Dye Yes
View More Specs

Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
15280114
View Documents
Thermo Scientific™
F175S
100 Reactions
119.00 €
100 reactions
Please to purchase this item. Need a web account? Register with us today!
15290114
View Documents
Thermo Scientific™
F175L
500 Reactions
514.00 €
500 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thermo Scientific Phusion Blood Direct PCR Master Mix is designed for amplification of DNA from whole blood. It eliminates the need for a separate DNA purification step prior to PCR. The modified Phusion Hot Start II High-Fidelity DNA Polymerase is resistant to main PCR inhibitors present in blood and retains its activity at blood concentrations of up to 40% in the PCR reaction.

This accurate proofreading DNA polymerase makes the Phusion Blood Direct PCR Master Mix an ideal choice for demanding applications, such as cloning. Phusion Blood Direct PCR Master Mix also allows analysis of blood samples with different preservatives (e.g., heparin, EDTA, citrate), storage conditions, and from various species.

Highlights

  • Direct PCR allows sample to be added directly to PCR reaction, reducing time-consuming and expensive DNA purification steps
  • Available Dilution & Storage protocol allows several PCR reactions from one sample
  • Specially engineered Phire Hot Start II DNA polymerase for extremely short PCR protocol times
  • Master mix format with premixed gel loading dye minimizes risk of cross-contamination, reduces sample handling, and allows direct loading on gel

Applications

  • Genotyping
  • Transgene detection
  • Knockout analysis
  • Sequencing
  • Cloning

Notes

  • Annealing temperature for Phire DNA Polymerases may differ significantly from that of Taq-based polymerases.
Specifications

Specifications

High
Phusion Blood II DNA Polymerase
Yes
Wet or Dry Ice
2X
Blunt
Blood
Direct PCR
Fast
Direct PCR Master Mix
Genotyping
Built-In Hot Start
SuperMix or Master Mix
DNA
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.