missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ultra-Sensitive ABC Peroxidase Standard Staining Kit

Detect biotinylated secondary antibodies by IHC using the highly sensitive avidin-biotin complex (ABC) technique.

Brand:  Thermo Scientific™ 32050

570.00 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 10188013

  • 616.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Pierce Ultrasensitive ABC Standard Peroxidase Staining Kit
4 mL
HRP (Horseradish Peroxidase) Substrate
Immunodetection, IHC (Immunohistochemistry)
Colorimetric, Biotin-Based, Chromogenic
  • Reagent A (Avidin), 2 mL
  • Reagent B (Biotinylated HRP), 2 mL
  • Mixing bottle with dropper dispenser tip, 1
  • Store all reagents at 4°C
Pierce™
Staining Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZI402934 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number32050
Lot NumberZH394326 Certificate TypeCertificate of Analysis Date04/09/2024 Catalog Number32050
Lot NumberYJ358663 Certificate TypeCertificate of Analysis Date09/11/2023 Catalog Number32050
Lot NumberYD358662 Certificate TypeCertificate of Analysis Date18/04/2023 Catalog Number32050
Lot NumberYA358661 Certificate TypeCertificate of Analysis Date02/02/2023 Catalog Number32050
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Ultra-Sensitive ABC Peroxidase Standard Staining Kit > Ultra-Sensitive ABC Standard Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.