missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ELISA Wash Buffer (30X)
Concentrated wash buffer for efficient use of storage in the lab
Brand: Thermo Scientific™ N503
This item is not returnable.
View return policy
Description
- Type: Reagent
- Format: 30X Concentrate

Specifications
Wash Buffer | |
Liquid | |
Store at -20°C. |
50 mL | |
ELISA |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ ELISA Wash Buffer (30X) > ELISA wash buffers; 30X concentrate
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction