Learn More
Thermo Scientific™ Pierce™ FlexMix™ Calibration Solution
Thermo Scientific™ Pierce™ FlexMix™ Calibration Solution is a mixture of 16 highly pure, ionizable components designed for both positive and negative ionization calibration using Orbitrap Tribrid Series 3.1 Instrument Control Software.
Brand: Thermo Scientific™ A39239
358.15 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
- Improved mass accuracy - extended mass range improves mass accuracy
- Positive and negative mode calibration - single formulation with extended mass ranges for both ionization calibration modes
- Highly validated - high-quality, rigorously-tested formulation with lot-specific certificates of analysis
- Ready-to-use - reference standard may be directly loaded into a syringe and injected into the instrument
- Store at room temperature for up to 18 months

Specifications
Mass Spectrometer | |
Amine | |
Room Temperature | |
10 mL |
Calibration Solution | |
Pierce™ | |
Calibration | |
Liquid |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Acetonitrile
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.