missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Glycoprotein Carbohydrate Estimation Kit

Measure protein glycosylation as the percent of purified protein mass compared to included glycoprotein standards; uses periodic acid Schiff-base reagents (PAS).

Brand:  Thermo Scientific™ 23260

Product Code. 10677914

  • 554.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Sufficient For: 60 tube assays or 250 microplate assays
• Sodium meta-Periodate, 500 mg
• Pierce Glycoprotein Detection Reagent, 500 mg
• Glycoprotein Assay Buffer, 250 mL
• Glycoprotein Standards Set, 5-protein set
Glycoprotein Carbohydrate Estimation
Pierce™
Not Target-Specific
Solution-based Detection, Absorbance
500 mg
Pierce Glycoprotein Carbohydrate Estimation Kit
Colorimetric
Glycoprotein Carbohydrate Estimation
60 Tube Assays or 250 Microplate Assays
Spectrophotometer, Microplate Reader
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZJ395886 Certificate TypeCertificate of Analysis Date08/11/2024 Catalog Number23260
Lot NumberZC386718 Certificate TypeCertificate of Analysis Date07/04/2024 Catalog Number23260
Lot NumberYJ372284 Certificate TypeCertificate of Analysis Date26/10/2023 Catalog Number23260
Lot NumberYF362995 Certificate TypeCertificate of Analysis Date09/06/2023 Catalog Number23260
Lot NumberXH352091 Certificate TypeCertificate of Analysis Date03/09/2022 Catalog Number23260
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Glycoprotein Carbohydrate Estimation Kit > Estimation Kit; 500mg kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.