Learn More
Thermo Scientific™ Guanidine-HCl
Pure, crystalline guanidium hydrochloride (GdnHCl, GuHCl), CAS 50-01-1; denaturant for protein biology methods (elution, solubilization, refolding, storage).
Brand: Thermo Scientific™ 24110
199.50 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Thermo Scientific™ Pierce Guanidine-HCl is pure, crystalline guanidine hydrochloride, a denaturant with many protein research applications.
Additional Information and Online Ordering
Highlights:
- Guanidine·HCl (Sequanal grade) is a highly purified, ready-to-use denaturant
- Free of UV-absorbing materials in the range of 225 – 300nm
- Sharp UV cut-off spectrum with A260 less than 0.03

Specifications
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C. | |
Chaotrope, Denaturant | |
Guanidine Hydrochloride |
Powder | |
500 g |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Guanidine Hydrochloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Guanidine hydrochloride
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.