missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein G Plus Agarose

High-capacity, recombinant Protein G beaded agarose resin for a variety of antibody affinity purification methods.

Brand:  Thermo Scientific™ 22851

 View more versions of this product

Product Code. 10220014

  • 498.00 € / 2mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Protein G Plus Agarose is a high-capacity Protein G beaded agarose resin for use in a variety of antibody affinity purification methods.

Pierce Protein G Plus Agarose consists of recombinant Protein G that has been covalently immobilized at high density onto high-quality crosslinked 6% beaded agarose (CL-6B). The resin is effective for affinity purification of IgG from serum and other fluids of many mammalian species. Protein G is especially well suited for use with mouse antibodies (including mouse IgG1) in addition to most IgG isotypes from human, goat and sheep samples.

Highlights:

  • Protein G – immobilized Protein G is ideal for polyclonal IgG purification from mouse, human, cow, goat and sheep serum, including human IgG3 and mouse IgG1 isotypes
  • Agarose resin – support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Inert and stable – superior manufacturing method immobilizes Protein G by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
  • High capacity – “Plus” variety of Pierce Protein G Agarose has a dense load of immobilized Protein G, providing a binding capacity greater than 20mg human IgG/mL resin

Properties of crosslinked 6% beaded agarose (CL-6B):

  • Support pH Stability: 2 to 14 (short term); 3 to 13 (long term)
  • Average Particle Size: 45 to 165 microns
  • Exclusion Limit: 10,000 to 4,000,000 daltons
  • Maximum Volumetric Flow Rate: approx. 1mL/minute (for 1cm diameter column)
  • Maximum Linear Velocity: 30cm per hour
  • Maximum Pressure: less than 25psi (1.5 bar), defined as the maximum pressure drop across a column that the resin can withstand (Note: The indicated gauge pressure of a liquid chromatography apparatus may be measuring the total system pressure rather than the pressure drop across the column.)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Agarose
Agarose Resin, Affinity
Protein G
Upon receipt store product at 4°C. Product is shipped at ambient temperature.
Pierce™
Bottle
IgG Antibodies
2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZI398115 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number22851
Lot NumberZI406464 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number22851
Lot NumberZC388583 Certificate TypeCertificate of Analysis Date17/03/2024 Catalog Number22851
Lot NumberYH378222 Certificate TypeCertificate of Analysis Date04/09/2023 Catalog Number22851
Lot NumberYB362923 Certificate TypeCertificate of Analysis Date09/03/2023 Catalog Number22851
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Protein G Plus Agarose > Resin; 2mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.