missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Maleimide Activated Horseradish Peroxidase Kit

Activated horseradish peroxidase (HRP) enzyme and kit to label proteins, peptides or other ligands via sulfhydryl groups (reduced cysteines) to make HRP conjugates.

Brand:  Thermo Scientific™ 31494

Product Code. 10454165

  • 504.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Maleimide-Activated HRP Conjugation Kit can be used to directly prepare HRP conjugates from proteins, peptides or other ligands that contain sulfhydryl groups, such as reduced cysteines.

The Thermo Scientific Maleimide Activated Horseradish Peroxidase (HRP) Kit can be used to directly prepare HRP conjugates from proteins, peptides or other ligands that contain a free sulfhydryl group. This kit eliminates the first step of the two-step maleimide method, making conjugate preparation much simpler. The kit is supplied with a stable, Maleimide-activated horseradish peroxidase. HRP conjugates can be purified by gel filtration chromatography, ultrafiltration or dialysis, depending upon the size of the conjugated protein.

Highlights:

  • Prepare enzyme conjugates from proteins that contain a free sulfhydryl
  • Includes 2-MEA to generate free sulfhydryls from disulfide bonds
  • Includes SATA to add free sulfhydryls to lysine residues
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Thiol
Maleimide
Sufficient For: 1 enzyme-labeling reaction with up to 5 mg of antibody (IgG)
• Maleimide Activated Horseradish Peroxidase, 5 mg
• Conjugation Buffer (10X), 20 mL
• PBS Pack (makes 500mL), 1 pack
• 2-Mercaptoethylamine-HCl, 6 mg
• SATA, 2 mg
• Dimethylformamide, 1 mL
• Hydroxylamine-HCl, 5 mg
• Polyacrylamide Desalting Column, 10 mL, 1 column

Store at 4°C.
EZ-Link™
Enzyme Labeled
1 Reaction
Water
Multi-component kit with activated HRP
1 Enzyme Labeling Reaction
HRP (Horseradish Peroxidase)
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZI400994 Certificate TypeCertificate of Analysis Date16/10/2024 Catalog Number31494
Lot NumberZI406423 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number31494
Lot NumberZE397117 Certificate TypeCertificate of Analysis Date04/06/2024 Catalog Number31494
Lot NumberZB394450 Certificate TypeCertificate of Analysis Date28/02/2024 Catalog Number31494
Lot NumberZA379912 Certificate TypeCertificate of Analysis Date06/02/2024 Catalog Number31494
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ EZ-Link™ Maleimide Activated Horseradish Peroxidase Kit > Maleimide Activated Hrp 1/pk

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.