Learn More
Pierce™ Pierce™ Ni-NTA Magnetic Agarose Beads
Thermo Scientific™ Pierce™ Ni-NTA Magnetic Agarose Beads provide a fast, convenient method for purification of His-tagged recombinant proteins.
324.00 € - 1204.00 €
Specifications
Concentration | Slurry: 25% |
---|---|
Shipping Condition | Wet Ice |
Particle Size | 30 μm |
Form | Liquid Suspension |
Product Line | Pierce™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15656144
|
Pierce™
78605 |
4 mL |
324.00 €
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15666144
|
Pierce™
78606 |
20 mL |
1204.00 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Product features:
Ni-NTA Magnetic Agarose Beads are used for small-scale affinity purification as well as high-throughput screening of recombinant His-tagged proteins. The polyhistidine tag is the most popular affinity tag and typically consists of six consecutive histidine residues (6xHis). Tagged proteins are overexpressed in a number of different systems, most commonly in bacteria, and purified from cell lysates such as those prepared using Thermo Scientific B-PER Bacterial Protein Extraction reagents. Purification of His-tagged proteins is achieved using an NTA chelate charged with nickel that coordinates with the histidine side chains. The NTA chelate contains four metal-binding sites that allow for low metal ion leaching and high binding capacity. The protocol for the Ni-NTA Magnetic Agarose Beads has been optimized to allow for high purity of the isolated His-tagged protein.
- High capacity—sufficient for both routine and demanding purification procedures; binds >70 mg of 6xHis-tagged protein per mL of settled resin
- High-performance and reproducible beads—non-aggregating, magnetite (Fe3O4), superparamagnetic beads provide exceptional uniformity for both manual and automated HTS purification applications
- Low nonspecific binding—optimized purification protocol results in better tagged-protein purification
- Versatile—purify proteins using native or denaturing conditions
- Compatible—use with Thermo Scientific Cell Lysis reagents and a variety of buffer additives
Applications:
- Protein purification and isolation
- High throughput protein purification using automated magnetic particle processor
Specifications
Slurry: 25% | |
30 μm | |
Pierce™ | |
Magnetic Agarose Bead | |
Bottle |
Wet Ice | |
Liquid Suspension | |
Recombinant | |
Metal Chelate, Divalent Nickel |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.