Learn More
Pierce™ Pierce™ Ni-NTA Magnetic Agarose Beads
Thermo Scientific™ Pierce™ Ni-NTA Magnetic Agarose Beads provide a fast, convenient method for purification of His-tagged recombinant proteins.
Brand: Pierce™ 78605
Description
Product features:
Ni-NTA Magnetic Agarose Beads are used for small-scale affinity purification as well as high-throughput screening of recombinant His-tagged proteins. The polyhistidine tag is the most popular affinity tag and typically consists of six consecutive histidine residues (6xHis). Tagged proteins are overexpressed in a number of different systems, most commonly in bacteria, and purified from cell lysates such as those prepared using Thermo Scientific B-PER Bacterial Protein Extraction reagents. Purification of His-tagged proteins is achieved using an NTA chelate charged with nickel that coordinates with the histidine side chains. The NTA chelate contains four metal-binding sites that allow for low metal ion leaching and high binding capacity. The protocol for the Ni-NTA Magnetic Agarose Beads has been optimized to allow for high purity of the isolated His-tagged protein.
- High capacity—sufficient for both routine and demanding purification procedures; binds >70 mg of 6xHis-tagged protein per mL of settled resin
- High-performance and reproducible beads—non-aggregating, magnetite (Fe3O4), superparamagnetic beads provide exceptional uniformity for both manual and automated HTS purification applications
- Low nonspecific binding—optimized purification protocol results in better tagged-protein purification
- Versatile—purify proteins using native or denaturing conditions
- Compatible—use with Thermo Scientific Cell Lysis reagents and a variety of buffer additives
Applications:
- Protein purification and isolation
- High throughput protein purification using automated magnetic particle processor

Specifications
1 mL | |
Slurry: 25% | |
Wet Ice | |
Liquid Suspension | |
4 mL | |
Magnetic Agarose Bead | |
Bottle |
4 mL | |
1 mL settled resin. Supplied as 4 mL of 25% v/v suspension in 20% ethanol. Store at 2–8°C. |
|
30 μm | |
Pierce™ | |
Recombinant | |
Metal Chelate, Divalent Nickel |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.