missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Pierce™ Peptide Retention Time Calibration Mixture Product Code.: 11572561
compliance-icons

Thermo Scientific™ Pierce™ Peptide Retention Time Calibration Mixture

Product Code. 11572561 Shop All Thermo Scientific Products
50 μL, 50µL
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
This item is not returnable. View return policy

Product Code. 11572561

Brand: Thermo Scientific™

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

15 synthetic heavy peptides mixed in equimolar ratio to test and calibrate reverse-phase liquid chromatography (LC) and mass spectrometry (LC-MS) equipment.

Thermo Scientific™ Pierce Peptide Retention Time Calibration Mixture enables reversed phase liquid chromatography (LC) and mass spectrometry (LC-MS) users to optimize and confirm correct operation of their equipment.

The Pierce Peptide Retention Time Calibration Mixture is used to optimize and assess LC parameters, identify total peptide elution window and optimize MS parameters. Testing the mixture frequently throughout the lifetime of a column or including it in quantitative samples makes it possible to monitor and normalize LC-MS performance between samples and over time. When used with Thermo Scientific Pinpoint 1.1 software, the calibration mixture is used to predict peptide retention times based upon peptide sequence and ease the transfer of methods between LC-MS platforms.

Highlights:

  • Assessment of chromatography and MS instrument performance
  • Prediction of peptide retention across multiple instrument platforms
  • Prediction of peptide retention time from sequence using calculated hydrophobicity factor
  • Optimization of scheduled MS acquisition windows for improved quantification and increased multiplexing
  • Internal standard to normalize for variation in retention times and peak intensities between runs
TRUSTED_SUSTAINABILITY

Specifications

Description Peptide Retention Time Calibration Mixture
Formulation Mixture of 15 synthetic heavy peptides at 0.5uM each
Product Type Peptide Retention Time Calibration Mixture
Sufficient For 5 to 100 uses
Quantity 50 μL
For Use With (Application) LC-MS
compliance-icons
Product Identifier
  • Pierce Peptide Retention Time Calibration Mixture
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : acetonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Peptide Retention Time Calibration Mixture >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.