missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Peroxidase Conjugate Stabilizer

An antifreeze solution to store aliquots and stocks of HRP-conjugated antibodies so they can be pipetted for use without thawing; better than glycerol.

Brand:  Thermo Scientific™ 31503

103.55 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.



Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11819160

  • 112.00 € / 25mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Peroxidase Conjugate Stabilizer is an antifreeze solution for storing aliquots and stocks of antibodies and other HRP-conjugates so they can be pipetted for use without thawing.

Highlights:

  • Provides a buffered antifreeze environment for enzyme conjugated antibodies and proteins Maintains a liquid, low viscosity solution at -20°C
  • No aliquoting of conjugate is necessary and sampling is convenient
  • Contains preservatives to prevent anti-microbial growth during long-term storage
TRUSTED_SUSTAINABILITY
Specifications

Specifications

25 mL
ELISA, Immunohistochemistry, Western Blot
Liquid
Stabilizing Reagent
Upon receipt store at 4°C.
Pierce™
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number091855 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number31503
Lot Number081455 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number31503
Lot Number011855 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number31503
Lot Number011655 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number31503
Lot Number051855 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number31503
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Peroxidase Conjugate Stabilizer > 25mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.