Learn More
Thermo Scientific™ PVDF Transfer Membranes, 0.45 μm, 10 Pre-cut Membranes
Pierce PVDF Transfer Membranes are made of high-quality polyvinylidene difluoride and provide high binding capacity for proteins and nucleic acids for western, Southern, and northern blotting methods.
Brand: Thermo Scientific™ 88585
196.65 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Pierce PVDF Transfer Membranes are made of high-quality polyvinylidene difluoride and provide high binding capacity for proteins and nucleic acids for western, Southern, and northern blotting methods. The 0.45-μm pore size make these membranes well-suited for a wide range of western blotting conditions and ideal for transfer of proteins >10 kDa. For best results, preactivate the membrane with 100% alcohol.
Features include:
• Re-probe characteristics—high mechanical strength makes PVDF an excellent membrane for stripping and re-probing
• Durability—compatible with most organic solvents, acids, and mild bases
• Compatible with commonly used transfer conditions and detection methods such as staining, chemiluminescence, and radiolabeling
• High sensitivity—exceptional binding capacity makes PVDF the ultimate choice for low abundance proteins

Specifications
10 cm | |
Pierce PVDF Transfer Membrane, 0.45 μm, 10 x 10 cm | |
Sheet | |
0.45 μm | |
Room Temperature | |
10 Mini-gel Blots |
10 cm | |
10 Pre-cut Membranes | |
PVDF | |
Store membranes flat at ambient temperature and away from chemical vapors. Some solvent vapors may partially dissolve the membranes, which will disrupt the pore structure. | |
10 x 10 cm |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ZJ397389 | Certificate of Analysis | 16/12/2024 | 88585_LITL18 PROMO,88585 |
ZI387418 | Certificate of Analysis | 15/10/2024 | 88585_LITL18 PROMO,88585 |
ZI387418A | Certificate of Analysis | 15/10/2024 | 88585_LITL18 PROMO,88585 |
ZF387417 | Certificate of Analysis | 07/08/2024 | 88585_LITL18 PROMO,88585 |
ZC387416 | Certificate of Analysis | 11/04/2024 | 88585_LITL18 PROMO,88585 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.