Learn More
Thermo Scientific™ TRACE™ TR-Wax GC Columns
Analyze a wide range of polar compounds with this low-bleed, general purpose, high polarity GC column.
Brand:  Thermo Scientific™ 260W131P
405.07 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10059142
  • 407.10 € / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Show More
Specifications
Specifications
260/280°C
0.32 mm
Trace™ TR-WAX
Ketones Analysis, GC for Alcohols, GC for Ketones, GC for Glycols, GC for Aromatic Isomers
15 m
15 m x 0.32 mm x 0.25 μm
0.32 mm
TRACE
G16, G20
0.25 μm
Polar
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ TRACE™ TR-Wax GC Columns >
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.