missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Rapid Antibody Isotyping Kit plus Kappa and Lambda - Mouse

Identify mouse monoclonal antibody class and subclass in 5 minutes with these single-use, cassette-based lateral-flow assays.

Brand:  Thermo Scientific™ 26179

365.75 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 10280434

  • 391.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Rapid Mouse Antibody Isotyping Kits are single-use, cassette-based kits for 5-minute lateral-flow assay determination of mouse monoclonal antibody class and subclass identity.

The kits are composed of small cartridges that provide a color-readout of the monoclonal antibody isotype within five minutes after pipetting a small amount of diluted culture supernatant or ascites fluid sample into the sample well. The kits determine mouse IgG1, IgG2a, IgG2b, IgG3, IgA, IgM.

The assay is performed by simply adding a properly diluted tissue culture supernatant or mouse ascites sample to the well of the small cassette. Gold conjugates embedded in the cassette form specific class- and subclass-soluble complexes with the antibodies in the sample. These complexes travel the length of the membrane and are resolved on the anti-isotype and class-specific antibody-impregnated membrane. Results are displayed as a red band indicating the antibody isotype or subclass.

Highlights:

  • Long shelf life – stable for more than 1 year at room temperature

  • Single-step – add diluted antibody sample to the loading-well of cassette

  • Fast – within 5 minutes a band appears that indicates the antibody isotype

  • Sensitive – detect antibodies in tissue culture media or ascites fluid samples at any concentration greater than 10ng/mL

  • Specific – assay does not crossreact with fetal bovine serum (FBS)

  • Reliable – results are similar to the standard ELISA-based isotyping assays

Includes:

Ten pouches, each containing two (Part No. 26178) or three (Part No. 26179) cassettes, plus a bottle of sample diluent

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pierce Rapid Isotyping Kit plus Kappa and Lambda - Mouse
Membrane Blot
Isotyping Assay
Pierce™
Sufficient For: 10 isotype determinations
• Isotyping Cassettes (three per pouch), 10 pouches
• Sample Diluent, 45 mL

Store kit at 2–8°C.
10 Tests
Chromogenic
Target-specific
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberSPECS Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number26179
Lot NumberZD386581 Certificate TypeCertificate of Analysis Date18/04/2024 Catalog Number26179
Lot NumberYJ358625 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number26179
Lot NumberXJ358624 Certificate TypeCertificate of Analysis Date11/11/2022 Catalog Number26179
Lot NumberXD330793 Certificate TypeCertificate of Analysis Date23/04/2022 Catalog Number26179
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Rapid Antibody Isotyping Kit plus Kappa and Lambda - Mouse > Kit for six isotypes, plus kappa and lambda identity; 10-test kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.