missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sulfo-NHS (N-hydroxysulfosuccinimide), No-Weigh™ Format

Prepare amine-reactive sulfo-NHS-ester crosslinkers or enhance carbodiimide (EDC) conjugation for peptides or proteins using N-hydroxysulfosuccinimide, CAS 106627-54-7.

148.00 € - 1842.00 €

Specifications

Molecular Weight (g/mol) 217.13
PEGylated No
Solubility Water
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10391314
View Documents
Thermo Scientific™
24510
500 mg
409.00 €
500mg
In Stock
to see stock.
Add to basket
 
11861365
View Documents
Thermo Scientific™
24520
8 x 2 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15993522
View Documents
Thermo Scientific™
A39269
10 x 2 mg
183.00 €
Pack of 10
In Stock
to see stock.
Add to basket
 
11871365
View Documents
Thermo Scientific™
24525
5 g
1842.00 €
5g
In Stock
to see stock.
Add to basket
 
Description

Description

Thermo Scientific™ Pierce Sulfo-NHS is pure N-hydroxysulfosuccinimide, the essential compound needed to prepare water-soluble, amine-reactive NHS-ester crosslinkers or to enhance conjugation efficiencies when using the carbodiimide EDC.

Highlights:

  • Forms an acyl amino ester with carboxyl groups when used with the EDC
  • Sulfo-NHS-activated molecule reacts efficiently with primary amines, resulting in stable amide bond
  • Allows for two-step EDC reactions between carboxyl- and amine-containing molecules
  • Sulfo-NHS-activated molecule can be stabilized by drying, allowing manufacture of amine-reactive probes and surfaces
  • Reactive groups: Sulfo-NHS
  • Reactive toward: amino groups (upon activation of carboxylates)
Specifications

Specifications

217.13
No
Water
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.