missing translation for 'onlineSavingsMsg'
Learn More
Piperonyl butoxide, 90%, Tech., Thermo Scientific Chemicals Product Code.: 10613352

Piperonyl butoxide, 90%, Tech., Thermo Scientific Chemicals

Product Code. 10613352
100 mL, Glass bottle
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10613352

Brand: Thermo Scientific Acros 334161000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-03-6
Molecular Formula C19H30O5
Molecular Weight (g/mol) 338.44
MDL Number MFCD00005842
InChI Key FIPWRIJSWJWJAI-UHFFFAOYSA-N
Synonym piperonyl butoxide, butacide, butocide, ethanol butoxide, pyrenone 606, piperonylbutoxide, 6-propylpiperonyl butyl diethylene glycol ether, butyl carbitol 6-propylpiperonyl ether, 5-2-2-butoxyethoxy ethoxy methyl-6-propylbenzo d 1,3 dioxole, butoxide synergistShow MoreShow Less
PubChem CID 5794
ChEBI CHEBI:32687
IUPAC Name 5-[2-(2-butoxyethoxy)ethoxymethyl]-6-propyl-1,3-benzodioxole
SMILES CCCCOCCOCCOCC1=CC2=C(OCO2)C=C1CCC

Specifications

Color Yellow
Density 1.0600g/mL
Boiling Point 180°C (1.0 mmHg)
Flash Point 170°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.4970 to 1.507
Quantity 100 mL
Beilstein 19, IV, 779
Merck Index 15, 7589
Specific Gravity 1.06
Solubility Information Solubility in water: practically insoluble. Other solubilities: miscible with methanol,ethanol,benzene,freons,,and other organic solvents
Viscosity 40 mPa.s (25°C)
Formula Weight 338.44
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Piperonyl butoxide, tech., 90%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0455637 Certificate TypeCertificate of Analysis Date29/08/2023 Catalog Number334165000,334161000
Lot NumberA0424486 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number334165000,334161000
Lot NumberA0400151 Certificate TypeCertificate of Analysis Date01/03/2021 Catalog Number334165000,334161000
compliance-icons compliance-icons
Product Identifier
  • Piperonyl butoxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit
Piperonyl butoxide, 90%, Tech., Thermo Scientific Chemicals >

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis