missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Indenti-plug™ Plastic Foam Stopper

Keep the contents of your containers secure with these durable and reusable stoppers. Indenti-plug™ Plastic Foam Stoppers can also be used as a cleaner for pipettes, tubes, vials and precision tubing.

Brand:  Cole-Parmer™ 06298-78

 View more versions of this product

Product Code. 11795299

  • 95.70 € / Pack of 100
Estimated Shipment: 23-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Breathable, nonabsorbent, chemical resistant and heat tolerant
  • Open-cell foam has a uniform density
  • Leaves no fibers
  • Autoclavable
Specifications

Specifications

35 to 45 mm
Autoclavable
Plastic
100/ck.
White
Plug Stopper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Indenti-plug™ Plastic Foam Stopper > Inner Diameter: 35 to 45mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.