missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Plastic Micropipet Stand
Designed for easy cleaning with a wide base and strong structure for great stability. Kartell™ Plastic Micropipet Stands hold 3 micropipets and can be sterilized by radiation.
Brand: Kartell™ 0131500
53.33 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
- Dimensions: 145W x 70mmH
- Convenient benchtop holder
Specifications
Transparent | |
Plastic | |
Micropipets | |
1 Pack |
Micropipettes | |
3 | |
1 Pack |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kartell™ Plastic Micropipet Stand > Holds: 3 Micropipettes
Spot an opportunity for improvement?Share a Content Correction