Learn More
KD Scientific Plastic Syringe
Economical and disposable, available in sizes ranging from 1 cc to 50 cc, Luer Slip (LS) or Luer Lock (LL) tip
Brand:  KD Scientific 780861
 View more versions of this product
Product Code. 15194204
Description
Description
  • A new line of disposable plastic sterile syringes for all scientific applications
  • Norm-ject syringes are the ideal solution for any situation
  • Their unique two-part system is latex free and contains no silicone lubricant or rubber
Specifications
Specifications
Polyethylene (Plunger)
Yes
Single-use
KD Scientific Syringe Pumps
Yes
No Needle Included
Syringe
30 mL
Luer Slip
Sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
KD Scientific Plastic Syringe > 30cc slip lock