Learn More
Thermo Scientific™ E1-ClipTip™ Bluetooth™ Electronic Multichannel Pipettes
GREENER_CHOICE
Automate daily 96-and 384-well microplate pipetting tasks with the sealed-in security of ClipTip technology in these electronic multichannel pipettes, which offer both versatility and customization.
Brand:  Thermo Scientific™ 4671100BT
922.35 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
922.35 EUR valid until 2025-05-31
Use promo code "25076" to get your promotional price.
 View more versions of this product
Product Code. 15613046
  • 1985.00 € / Each
Explore more special offers
Description
Show More
Specifications
Specifications
ClipTip 1250
ClipTip™ 1250
8
Yes
15 to 1250 μL
Turquoise
Bluetooth
E1-ClipTip
Electronic Pipette
1μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ E1-ClipTip™ Bluetooth™ Electronic Multichannel Pipettes > 8-channel, 15 to 1250μL