Learn More
Thermo Scientific™ PocketTip™ D.A.R.T.'S™ (Disposable Automation Research Tips)
Reduce reagent, labware and sample costs while improving data quality through the elimination of intermediate pipetting steps with soft in-tip mixing.
Brand: Thermo Scientific™ 5696-50
Description
Reduce reagent, labware and sample costs while improving data quality through the elimination of intermediate pipetting steps with soft in-tip mixing. Thermo Scientific™ PocketTip™ D.A.R.T.'s™ achieve lower volume transfer capability due to a combination of the internally molded capillary pocket and the unique surface sealing of the tip.
Features:
- Allows existing Thermo Scientific™ Matrix™ automated liquid handlers to accommodate low-volume dispensing for compound screening procedures
- Requires only minor procedural adjustments to Thermo Scientific™ PlateMate™ and Thermo Scientific™ Hydra™ DT instrumentation
- Eliminate aqueous dilution steps that can cause precipitation by transferring nanoliter volumes directly to assay plate.
- Control consistent accuracy and precision during the molding process
- Purity and straightness validation are performed on compatible instrumentation to ensure only the highest quality.
- With instrument compatibility range starting from the Matrix Hydra DT, on-the-fly dose response curves or assay development can be set up with simple Hydra DT programming.
- Higher throughput needs can be met with the Matrix PlateMate 2 x 3 and Catalyst Express or Matrix PlateMate Plus instruments.
- Sales specialists are available to assist with instrument programming and modifications or instrument validation of any Matrix automated liquid handler.
- Available in 96- and 384- channel format with 50, 100 and 250nL pocket sizes
- Sterile format available for sensitive liquid handling procedures
Specifications
50 nL | |
Non-sterile | |
Automation | |
Single-use | |
PocketTip |
Non-filtered | |
Rack | |
20 x 96 tips | |
Thermo Scientific Matrix Hydra DT, Thermo Scientific PlateMate Plus |
Product Suggestions
Customers who viewed this item also viewed
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.