Learn More
Eppendorf epT.I.P.S.™ Motion Pipette Tips
GREENER_CHOICE
Eppendorf Totally Integrated Pipetting System for Automation. Works in perfect combination with epMotion platform.
Brand:  Eppendorf 0030015258
255.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 30019041
  • 259.00 € / Pack of 960
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Specially designed for use on the epMotion™ 5070, 5073 and 5075 Liquid Handling Workstation
  • Motion pipette tips, with and without filters, developed in torsion-resistant, clean racks
  • High-precision manufacturing guarantees a perfect fit and extremely accurate dispensing behavior
  • Optimized for simple automatic ejection in the Liquid Handling Workstation collection vessel
  • Can also be integrated into the workstation as a refilling system
  • Packaged in contamination-free racks
  • Available in the following purity grades: standard Eppendorf Quality, PCR clean, Sterile, PCR clean and Sterile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1000 μL
Sterile
Not Autoclavable
Box
Robotic Tips
Eppendorf epMotion
Polypropylene
Filtered
Clear
For Eppendorf epMotion series
Standard, Straight
Single-use
No
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf epT.I.P.S.™ Motion Pipette Tips > Volume (Metric): 1000 μL; Filtered: Filtered; Sterility: Sterile