missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(vinyl chloride), carboxylated, aver. M.W.220000, aver. carboxyl content 1.8 wt.%, Thermo Scientific Chemicals
71.76 € - 141.71 €
Chemical Identifiers
CAS | 25702-80-1 |
---|---|
Molecular Formula | (C2H3R)n |
Molecular Weight (g/mol) | 134.56 |
MDL Number | MFCD00084460 |
InChI Key | SQNNHEYXAJPPKH-UHFFFAOYSA-N |
Synonym | 2-propenoic acid, polymer with chloroethene, polyvinyl chloride-acrylic copolymer, acrylic acid; chloroethylene, chloroethene; prop-2-enoic acid, acrylic acid; vinyl chloride |
PubChem CID | 3080734 |
IUPAC Name | chloroethene; prop-2-enoic acid |
SMILES | *-CC(-*)[*] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10573741
|
Thermo Scientific Acros
183311000 |
100 g | Glass bottle |
71.76 €
100g |
In Stock
Log in to see stock. |
||||
10183960
|
Thermo Scientific Acros
183312500 |
250 g | Glass bottle |
141.71 €
250g |
Estimated Shipment: 26-11-2024
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25702-80-1 | |
134.56 | |
SQNNHEYXAJPPKH-UHFFFAOYSA-N | |
3080734 | |
*-CC(-*)[*] |
(C2H3R)n | |
MFCD00084460 | |
2-propenoic acid, polymer with chloroethene, polyvinyl chloride-acrylic copolymer, acrylic acid; chloroethylene, chloroethene; prop-2-enoic acid, acrylic acid; vinyl chloride | |
chloroethene; prop-2-enoic acid |
Specifications
White | |
Amorphous Powder | |
MFCD00084460 | |
2-propenoic acid, polymer with chloroethene, polyvinyl chloride-acrylic copolymer, acrylic acid; chloroethylene, chloroethene; prop-2-enoic acid, acrylic acid; vinyl chloride | |
*-CC(-*)[*] | |
134.56 | |
Authentic | |
Poly(vinyl chloride), carboxylated |
25702-80-1 | |
(C2H3R)n | |
1.3900g/mL | |
SQNNHEYXAJPPKH-UHFFFAOYSA-N | |
chloroethene; prop-2-enoic acid | |
3080734 | |
1.39 |
Safety and Handling
Product Identifier
- Poly(vinyl chloride), carboxylated, aver. M.W.220000, aver. carboxyl content 1.8 wt%
RUO – Research Use Only