Learn More
Product Code | Brand | dimCapacityMetric | dimClosureType | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | dimCapacityMetric | dimClosureType | Price | Quantity & Availability | |||||
11934385
|
Fisherbrand™
14955191 |
81.00 €
Pack of 250 |
Estimated Shipment: 26-09-2025
Log in to see stock. |
|||||||
11984385
|
Fisherbrand™
14955194 |
90.00 €
Pack of 250 |
Estimated Shipment: 26-09-2025
Log in to see stock. |
|||||||
11974395
|
Fisherbrand™
14955185 |
120.00 €
Pack of 500 |
Estimated Shipment: 26-09-2025
Log in to see stock. |
|||||||
11944405
|
Fisherbrand™
14955182 |
73.80 €
Pack of 500 |
In Stock
Log in to see stock. |
|||||||
11984395
|
Fisherbrand™
14955189 |
104.00 €
Pack of 500 |
Estimated Shipment: 26-09-2025
Log in to see stock. |
|||||||
11954405
|
Fisherbrand™
14955181 |
144.00 €
Pack of 500 |
Estimated Shipment: 26-09-2025
Log in to see stock. |
|||||||
11974405
|
Fisherbrand™
14955188 |
135.00 €
Pack of 500 |
Estimated Shipment: 26-09-2025
Log in to see stock. |
|||||||
Description
- Secure, contaminant free bags to ensure dependable analysis results
- Made from heavy gauge, virgin polyethylene that meets FDA specifications
- Can accommodate solid, semi solid and liquid materials
- Easy to use, insert sample and seal by twirling the bag away outward four times and bending wire ends inward
- Economical and efficient way to collect, contain and carry samples
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Polyethylene Sample Bags