Learn More
Fisherbrand™ Polyethylene Sample Bags
For use in the laboratory to carry samples into a sterile environment.
Brand:  Fisherbrand™ 14955181
 View more versions of this product
Product Code. 11954405
  • 144.00 € / Pack of 500
Estimated Shipment: 01-10-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Secure, contaminant free bags to ensure dependable analysis results
  • Made from heavy gauge, virgin polyethylene that meets FDA specifications
  • Can accommodate solid, semi solid and liquid materials
  • Easy to use, insert sample and seal by twirling the bag away outward four times and bending wire ends inward
  • Economical and efficient way to collect, contain and carry samples
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Polyethylene Sample Bags > Capacity: 1.26L; Round wire closure