Learn More
Fisherbrand™ Polypropylene Pipet Finntip™
Make pipetting simple and safer for even the most sensitive applications. Fisherbrand™ Polypropylene Pipet Finntip™s are perfectly fit for manual or electronic, single or multichannel pipets.
Brand:  Fisherbrand™ FB34661
 View more versions of this product
Product Code. 11527462
  • 115.00 € / Pack of 250
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • Practice-oriented tip volumes
  • Aeroject™ tips with special filters made of hydrophobic PE-HD as
  • Aerosol barrier and for protection against contamination
  • Suitable for all commonly used pipet types
Specifications
Specifications
1000 to 5000 μL
Natural
FinnTip
Non-sterile
Autoclavable
Polypropylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Polypropylene Pipet Finntip™ > Capacity: 1000 to 5000uL