Learn More
Eppendorf™ Polypropylene V-Bottom 384 Well Microplate
Brand: Eppendorf™ 0030621670
890.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Specifications
PCR clean | |
16 x 24 | |
Gray, White Bottom, White Bottom | |
384 | |
80/Pk. |
Standard | |
Autoclavable | |
Polypropylene | |
Conical Bottom (V-well) |