Learn More
Eppendorf™ Polypropylene V-Bottom 384 Well Microplate
Brand: Eppendorf™ 0030625307
1396.00 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
PCR clean | |
16 x 24 | |
White, Clear Bottom, Clear Bottom | |
384 | |
240/Pk. |
Standard | |
Autoclavable | |
Polypropylene | |
Conical Bottom (V-well) |