Learn More
Fisherbrand™ Polypropylene Yellow Pipet Tip E
Make pipetting simple and safer for even the most sensitive applications. Fisherbrand™ Polypropylene Yellow Pipet Tips E are perfectly fit for manual or electronic, single or multichannel pipets.
Brand:  Fisherbrand™ FB34531
Product Code. 11587442
  • 39.25 € / Pack of 1000
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Available pre-sterilized or non-sterile
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • Practice-oriented tip volumes
  • Aeroject™ tips with special filters made of hydrophobic PE-HD as
  • Aerosol barrier and for protection against contamination
  • Suitable for all commonly used pipet types
Specifications
Specifications
1 to 200 μL
Yellow
Autoclavable
Polypropylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Polypropylene Yellow Pipet Tip E > Volume Metric: 1 to 200μL