Learn More
Fisherbrand™ Polystyrene Macro Cuvettes for UV and Visible Wavelengths
Description
Cuvets with improved photometric properties.- Very low variation of extinction values
- Excellent optical transmission range
Specifications
Macro Cuvette | |
4 mL | |
100/Pk. | |
45 mm |
Reusable | |
Plastic | |
220 to 900 nm | |
12.5 mm |
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Product Title
Fisherbrand™ Polystyrene Macro Cuvettes for UV and Visible Wavelengths > Capacity: 4mL; Wavelength: 220 to 900nm; QTY: 100/Pk.
Spot an opportunity for improvement?Share a Content Correction