missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polystyrene Macro Cuvettes for Visible Wavelengths

Use these cuvettes for more constant sample temperatures during photometric measurements. Fisherbrand™ Polystyrene Macro Cuvettes for Visible Wavelengths feature excellent optical transmission range and small wall thickness for increased heat transfer.

Brand:  Fisherbrand™ FB55143

Product Code. 11537692

  • 13.45 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Very low variation of extinction values
  • Excellent optical transmission range
  • Cavity-sorted production
  • Transparency Approximately 90% off between 400nm and 800nm
Specifications

Specifications

Macro Cuvette
4 mL
100/Pk.
400 to 800 nm
12.5 mm
Single-use
Polystyrene
10 mm
45 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Polystyrene Macro Cuvettes for Visible Wavelengths > Capacity: 4mL; Wavelength: 400 to 800nm; QTY: 100/Pk.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.