Learn More
LABPLAS Polyurethane Sani-Stick with Dey-Engley Neutralizing Broth
Synthetic polyurethane sponge with stick for sampling surfaces, with buffer to neutralize disinfectant.
Brand:  LABPLAS KSS-67915-DE
 View more versions of this product
Product Code. 17936649
  • 326.00 € / Pack of 100
Explore more special offers
Description
Description
  • Polyurethane Sani-Stick
  • Saturated with Dey-Engley neutralizing broth
  • Dimensions: 160 x 229 mm
  • Holds: 600 mL
  • Option to include gloves with kit
Specifications
Specifications
Hygiene Sponge Sampling Kit
Neutralizers
Sterile
Gloves
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
LABPLAS Polyurethane Sani-Stick with Dey-Engley Neutralizing Broth > Gloves