missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide, ACS Reagent, pellets, ≥85%, Honeywell Fluka™
Product Code.: 15672140
- Home
- Products
- Chemicals
- Bases
- Potassium Hydroxide
- Potassium hydroxide, ACS Reagent, pellets, ≥85%, Honeywell Fluka™
Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
Potassium hydroxide, ACS Reagent, pellets, ≥85%, Honeywell Fluka™
Product Code. 156721406 x 500 g
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
6 x 500 g
12 kg
This item is not returnable.
View return policy
Product Code. 15672140
Brand: Honeywell-Fluka 2214736X500G
This item is not returnable.
View return policy
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Chemical Identifiers
CAS | 1310-58-3 |
Molecular Formula | HKO |
Molecular Weight (g/mol) | 56.11 |
MDL Number | MFCD00003553 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693Show MoreShow Less |
PubChem CID | 14797 |
ChEBI | CHEBI:32035 |
SMILES | [OH-].[K+] |
Specifications
Linear Formula | KOH |
UN Number | 1813 |
Quantity | 6 x 500 g |
IUPAC Name | potassium;hydroxide |
Formula Weight | 56.11g/mol |
Percent Purity | ≥85% |
Grade | ACS Reagent |
Vapor Pressure | 1mmHg (719°C) |
Chemical Name or Material | Potassium Hydroxide |
Product Suggestions
You may be interested in these alternatives
-
Product Code. 10142010
-
Product Code. 10474321
-
Product Code. 11336808
-
Product Code. 15663070
-
Product Code. 15622140
Customers who viewed this item also viewed This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
Viewing 1-5 of 12
-
Catalog No. 10316380
67,75 € / 2.50 L -
Catalog No. 10596814
90,45 € / 50mL -
Catalog No. 10304980
160,00 € / 2.50 L -
Catalog No. 10793611
154,00 € / 1kg -
Catalog No. 10112640
33,95 € / 1kg -
Catalog No. 10092740
27,44 € / 1kg -
Catalog No. 11448842
43,35 € / 2.50 L -
Catalog No. 10375810
63,25 € / 500g -
Catalog No. 10117113
34,57 € / 100g -
Catalog No. 10448990
63,15 € / 1kg -
Catalog No. 10509390
30,60 € / 500g -
Catalog No. 10113190
47,44 € / 1kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium hydroxide, ACS Reagent, pellets, ≥85%, Honeywell Fluka™ >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Spot an opportunity for improvement?Share a Content Correction