missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Potassium permanganate, 98%, extra pure Product Code.: 10092610

Thermo Scientific Chemicals Potassium permanganate, 98%, extra pure

Product Code. 10092610
1 kg
Click to view available options
Quantity:
25 g
250 g
500 g
1 kg
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Product Code. 10092610

Brand: Thermo Scientific Chemicals 196750010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-64-7
Molecular Formula KMnO4
MDL Number MFCD00011364
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san sooShow MoreShow Less
PubChem CID 516875

Specifications

Color Brown to Black
Physical Form Lustrous Crystalline Powder
Quantity 1 kg
Assay Percent Range 98%
Linear Formula KMnO4
Fieser 01,942; 02,348; 04,412; 05,562; 08,416; 09,388; 10,330; 11,440; 12,413; 13,258; 14,267; 15,273; 16,283
Merck Index 15, 7776
SMILES [O-][Mn](=O)(=O)=O.[K+]
IUPAC Name potassium;permanganate
Molecular Weight (g/mol) 158.032
Formula Weight 158.04
Percent Purity ≥97.5%
Grade Extra Pure
Packaging Plastic bottle
Density 2.7
Chemical Name or Material Potassium permanganate, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468123 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number196750010,196752500,196755000,196750050
Lot NumberA0468122 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number196750010,196752500,196755000,196750050
Lot NumberA0464338 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog Number196750010,196752500,196755000,196750050
Lot NumberA0459166 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog Number196750250,196750010,196752500,196755000,196750050
Lot NumberA0450403 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number196750010,196752500,196755000,196750050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Potassium permanganate, 98%, extra pure >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.