missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PQ 401, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Insulin-like growth factor receptor (IGF1R) inhibitor. Suppresses IGF-stimulated IGF-IR autophosphorylation with an IC50 value of 12 μM. Inhibits growth of MCF-7 breast cancer cells in vitro and in vivo.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 341.79 |
Percent Purity | >99% |
Chemical Name or Material | PQ 401 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'